5-(Ethylthio)-1H-tetrazole (ETT) ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Ethyltio-1H-Tetrazole

003950-25G 003950-100G
101786-508EA 115.38 USD
101786-508 101786-510
5-(Ethylthio)-1H-tetrazole (ETT) ≥97%
5-(Ethylthio)-1H-tetrazole (ETT)

Hazard: Irritant

Formula: C₃H₆N₄S
MW: 130.17 g/mol
Melting Pt: 88…89 °C
MDL Number: MFCD00068733
CAS Number: 89797-68-2

Order Now

Specification Test Results

Melting point 88-89°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR