1-Iodo-4-phenoxybenzene ≥98%
Supplier: Matrix Scientific
003868-5G
003868-25G
101786-338EA
75.38
USD
101786-338
101786-340
1-Iodo-4-phenoxybenzene ≥98%
1-Iodo-4-phenoxybenzene
Hazard: Irritant
Formula:
C₁₂H₉IO MW: 296.1037 g/mol Melting Pt: 144…146 °C |
MDL Number:
MFCD00060666 CAS Number: 2974-94-9 |
Specification Test Results
Melting point | 144-146°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...