3,3,3-Trifluoropropan-1-amine hydrochloride ≥97%

Supplier: Matrix Scientific

003811-250MG 003811-1G 003811-5G
101786-280EA 30.77 USD
101786-280 101786-282 101786-284
3,3,3-Trifluoropropan-1-amine hydrochloride ≥97%
3,3,3-Trifluoropropan-1-amine hydrochloride

Hazard: Irritant

Formula: C3H7ClF3N
Melting Pt: 222…223 °C
MDL Number: MFCD00236725
CAS Number: 2968-33-4

Order Now

Specification Test Results

Melting point 222-223°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR