1-(2,4-Difluorophenyl)biguanide hydrochloride

Supplier: Matrix Scientific

003800-1G 003800-5G
101786-264EA 30.77 USD
101786-264 101786-266
1-(2,4-Difluorophenyl)biguanide hydrochloride
1-(2,4-Difluorophenyl)biguanide hydrochloride

Hazard: Irritant

Formula: C₈H₁₀ClF₂N₅
Melting Pt: 226-228°C
MDL Number: MFCD00053015
CAS Number: 66088-52-6

Order Now

Specification Test Results

Melting point 226-228°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR