3,4-Difluoro-DL-phenylalanine ≥97%

Supplier: Matrix Scientific

003783-1G 003783-5G
101786-230EA 235.38 USD
101786-230 101786-232
3,4-Difluoro-DL-phenylalanine ≥97%
3,4-Difluoro-DL-phenylalanine

Hazard: Irritant

Formula: C₉H₉F₂NO₂
Melting Pt: 237…240 °C
MDL Number: MFCD00061307
CAS Number: 32133-36-1

Order Now

Specification Test Results

Melting point 237-240°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR