N-[4-(Trifluoromethyl)phenyl]-4-(trifluoromethyl)benzamide ≥97%

Supplier: Matrix Scientific

003749-1G 003749-5G
101786-160EA 103.08 USD
101786-160 101786-162
N-[4-(Trifluoromethyl)phenyl]-4-(trifluoromethyl)benzamide ≥97%
N-[4-(Trifluoromethyl)phenyl]-4-(trifluoromethyl)benzamide

Hazard: Irritant

Formula: C₁₅H₉F₆NO
MW: 333.24 g/mol
Melting Pt: 206…207 °C
MDL Number: MFCD00045101
CAS Number: 195371-90-5

Order Now

Specification Test Results

Melting point 206-207°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR