3,3,3-Trifluoro-DL-alanine ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Amino-3,3,3-trifluoropropanoic acid, H-DL-3,3,3-TriF-Ala-OH
003593-250MG
003593-1G
101785-896EA
130.77
USD
101785-896
101785-898
3,3,3-Trifluoro-DL-alanine ≥97%
3,3,3-Trifluoro-DL-alanine
Hazard: Irritant
Formula:
C₃H₄F₃NO₂ Melting Pt: 232…234 °C |
MDL Number:
MFCD00004263 CAS Number: 17463-43-3 |
Specification Test Results
Melting point | 232-234°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...