1-(2-Naphthoyl)-3,3,3-trifluoroacetone ≥98%

Supplier: Matrix Scientific

Synonyms: 4,4,4-Trifluoro-1-(naphthalen-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione

003550-5G 003550-25G
101785-854EA 29.23 USD
101785-854 101785-856
1-(2-Naphthoyl)-3,3,3-trifluoroacetone ≥98%
1-(2-Naphthoyl)-3,3,3-trifluoroacetone

TCA listed: T
Hazard: Irritant

Formula: C₁₄H₉F₃O₂
MW: 266.22 g/mol
Melting Pt: 74-76°C
MDL Number: MFCD00054501
CAS Number: 893-33-4

Order Now

Specification Test Results

Melting point 74-76°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR