3,3-Bis(methylthio)-2-[3-(trifluoromethyl)benzoyl]acrylonitrile
Supplier: Matrix Scientific
003417-250MG
003417-1G
101785-674EA
55.38
USD
101785-674
101785-676
3,3-Bis(methylthio)-2-[3-(trifluoromethyl)benzoyl]acrylonitrile
3,3-Bis(methylthio)-2-[3-(trifluoromethyl)benzoyl]acrylonitrile
Hazard: Irritant
Formula:
C₁₃H₁₀F₃NOS₂ Melting Pt: 60 °C |
MDL Number:
MFCD00174303 CAS Number: 116492-97-8 |
Specification Test Results
Melting point | 60°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...