1,4-Bis(tert-butoxy)tetrafluorobenzene ≥90%, Technical Grade

Supplier: Matrix Scientific

003363-5G 003363-25G
101785-610EA 66.15 USD
101785-610 101785-612
1,4-Bis(tert-butoxy)tetrafluorobenzene ≥90%, Technical Grade
1,4-Bis(tert-butoxy)tetrafluorobenzene

Hazard: Irritant

Formula: C₁₄H₁₈F₄O₂
Melting Pt: 55…56 °C
MDL Number: MFCD00069107
CAS Number: 121088-09-3

Order Now

Specification Test Results

Melting point 55-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR