5-Chlorobenzofuran-2-carboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Chlorobenzo[b]furan-2-carboxylic acid
003287-1G
003287-5G
101785-512EA
36.92
USD
101785-512
101785-514
5-Chlorobenzofuran-2-carboxylic acid ≥97%
5-Chlorobenzofuran-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₅ClO₃ MW: 196.59 g/mol Melting Pt: ~275°C (sub) |
MDL Number:
MFCD00060512 CAS Number: 10242-10-1 |
Specification Test Results
Melting point | ~275°C(sub) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...