4-Nitrophenyl phenyl sulfide
Supplier: Matrix Scientific
Synonyms:
(4-Nitrophenyl)(phenyl)sulphane
003219-5G
003219-25G
101785-418EA
52.31
USD
101785-418
101785-420
4-Nitrophenyl phenyl sulfide
4-Nitrophenyl phenyl sulphide
Hazard: Irritant
Formula:
C₁₂H₉NO₂S MW: 231.27 g/mol Melting Pt: 56-58°C |
MDL Number:
MFCD00024700 CAS Number: 952-97-6 |
Specification Test Results
Melting point | 56-58°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...