4-Nitrophenyl anthranilate ≥98%
Supplier: Matrix Scientific
003201-5G
003201-25G
101785-402EA
180
USD
101785-402
101785-404
4-Nitrophenyl anthranilate ≥98%
4-Nitrophenyl anthranilate
Hazard: Irritant
Formula:
C₁₃H₁₀N₂O₄ Melting Pt: 127…129 °C |
MDL Number:
MFCD00056116 CAS Number: 19176-60-4 |
Specification Test Results
Melting point | 127-129°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...