(Pentafluorophenyl)diphenylphosphine ≥97%

Supplier: Matrix Scientific

Synonyms: Diphenyl(pentafluorophenyl)phosphine

003191-1G 003191-5G
101785-386EA 407.69 USD
101785-386 101785-388
(Pentafluorophenyl)diphenylphosphine ≥97%
(Pentafluorophenyl)diphenylphosphine

Hazard: Irritant

Formula: C₁₈H₁₀F₅P
MW: 352.24 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD00000290
CAS Number: 5525-95-1

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR