5-(4-Pyridyl)-1H-tetrazole ≥98%
Supplier: Matrix Scientific
Synonyms:
4-(1H-Tetrazol-5-yl)pyridine
003162-1G
003162-5G
101785-348EA
76.92
USD
101785-348
101785-350
5-(4-Pyridyl)-1H-tetrazole ≥98%
5-(4-Pyridyl)-1H-tetrazole
Hazard: Irritant-Harmful
Formula:
C₆H₅N₅ MW: 147.14 Melting Pt: 265…267 °C |
MDL Number:
MFCD00068115 CAS Number: 14389-12-9 |
Specification Test Results
Melting point | 265-267°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...