5-(3-Pyridyl)-1H-tetrazole ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(2H-Tetrazol-5-yl)pyridine

003161-1G 003161-5G
101785-344EA 76.92 USD
101785-344 101785-346
5-(3-Pyridyl)-1H-tetrazole ≥98%
5-(3-Pyridyl)-1H-tetrazole

Hazard: Irritant-Harmful

Formula: C₆H₅N₅
MW: 147.14 g/mol
Melting Pt: 240…242 °C
MDL Number: MFCD00040281
CAS Number: 3250-74-6

Order Now

Specification Test Results

Melting point 240-242°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR