4-Acetamidophenylacetonitrile

Supplier: Matrix Scientific

Synonyms: 4'-(Cyanomethyl)acetanilide, N-(4-(Cyanomethyl)phenyl)acetamide

003156-1G 003156-5G
101785-330EA 36.92 USD
101785-330 101785-332
4-Acetamidophenylacetonitrile
4-Acetamidophenylacetonitrile

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
Melting Pt: 82…83 °C
MDL Number: MFCD00016398
CAS Number: 25025-06-3

Order Now

Specification Test Results

Melting point 82-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR