Malonic acid dihydrazide

Supplier: Matrix Scientific

003109-5G 003109-25G
101785-250EA 63.08 USD
101785-250 101785-252
Malonic acid dihydrazide
Malonic acid dihydrazide

Hazard: Irritant

Formula: C₃H₈N₄O₂
Melting Pt: 157…159 °C
MDL Number: MFCD00041268
CAS Number: 3815-86-9

Order Now

Specification Test Results

Melting point 157-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR