Methyl-4-chloro-6-(trifluoromethyl)thieno[3,4-B]pyridine-1-carboxylate

Supplier: Matrix Scientific

003090-250MG 003090-1G
101785-208EA 56.92 USD
101785-208 101785-210
Methyl-4-chloro-6-(trifluoromethyl)thieno[3,4-B]pyridine-1-carboxylate
Methyl-4-chloro-6-(trifluoromethyl)thieno[3,4-B]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₅ClF₃NO₂S
Melting Pt: 103…106 °C
MDL Number: MFCD00173840
CAS Number: 175203-40-4

Order Now

Specification Test Results

Melting point 103-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR