4-Phenoxybenzonitrile ≥98%
Supplier: Matrix Scientific
003020-5G
003020-25G
101785-094EA
36.92
USD
101785-094
101785-096
4-Phenoxybenzonitrile ≥98%
4-Phenoxybenzonitrile
Hazard: Irritant
Formula:
C₁₃H₉NO MW: 195.22 g/mol Melting Pt: 44-46°C |
MDL Number:
MFCD00017346 CAS Number: 3096-81-9 UN: 3439 ADR: 6.1,III |
Specification Test Results
Melting point | 44-46°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...