Ethyl-7-methoxybenzofuran-2-carboxylat ≥98%
Supplier: Matrix Scientific
Synonyms:
Ethyl 7-methoxybenzofuran-2-carboxylate
003016-1G
003016-5G
101785-082EA
29.23
USD
101785-082
101785-084
Ethyl-7-methoxybenzofuran-2-carboxylat ≥98%
Ethyl 7-methoxybenzofuran-2-carboxylat
Hazard: Irritant
Formula:
C₁₂H₁₂O₄ MW: 220.22 g/mol Melting Pt: 87-89°C |
MDL Number:
MFCD00060510 CAS Number: 50551-58-1 |
Specification Test Results
Melting point | 87-89°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...