5-Methoxybenzofuran-2-carboxylic acid ≥98%

Supplier: Matrix Scientific

003013-1G 003013-5G
101785-070EA 56.92 USD
101785-070 101785-072
5-Methoxybenzofuran-2-carboxylic acid ≥98%
5-Methoxybenzofuran-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈O₄
MW: 192.1700 g/mol
Melting Pt: 212-214°C
MDL Number: MFCD00079771
CAS Number: 10242-08-7

Order Now

Specification Test Results

Melting point 212-214°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR