Mandelic acid hydrazide ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Hydroxy-2-phenylacetohydrazide
002932-5G
002932-25G
101784-946EA
46.15
USD
101784-946
101784-948
Mandelic acid hydrazide ≥97%
Mandelic acid hydrazide
TCA listed: T
Hazard: Irritant
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol Melting Pt: 134…136 °C |
MDL Number:
MFCD00038133 CAS Number: 2443-66-5 |
Specification Test Results
Melting point | 134-136°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...