2-Chloro-N-[4-(trifluoromethyl)phenyl]acetamide
Supplier: Matrix Scientific
Synonyms:
N-(Chloroacetyl)-4-(trifluoromethyl)aniline, 2-Chloro-N-(4-(trifluoromethyl)phenyl)acetamide
002930-5G
002930-25G
101784-942EA
43.08
USD
101784-942
101784-944
2-Chloro-N-[4-(trifluoromethyl)phenyl]acetamide
2-Chloro-N-[4-(trifluoromethyl)phenyl]acetamide
Hazard: Irritant
Formula:
C₉H₇ClF₃NO MW: 237.61 g/mol Melting Pt: 158…161 °C |
MDL Number:
MFCD00084933 CAS Number: 2707-23-5 |
Specification Test Results
Melting point | 158-161°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...