N-(Chloroacetyl)-3-(trifluoromethyl)aniline ≥97%

Supplier: Matrix Scientific

002927-5G 002927-25G
101784-932EA 38.46 USD
101784-932 101784-934
N-(Chloroacetyl)-3-(trifluoromethyl)aniline ≥97%
N-(Chloroacetyl)-3-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₉H₇ClF₃NO
MW: 237.606 g/mol
Melting Pt: 62.5…64 °C
CAS Number: 351-38-2

Order Now

Specification Test Results

Melting point 62.5-64°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR