2-Chloro-2'-(trifluoromethyl)acetanilide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-N-(2-(trifluoromethyl)phenyl)acetamide, N-Chloroacetyl-2-(trifluoromethyl)aniline

002925-1G 002925-5G
101784-922EA 49.23 USD
101784-922 101784-924
2-Chloro-2'-(trifluoromethyl)acetanilide ≥97%
2-Chloro-2'-(trifluoromethyl)acetanilide

Hazard: Irritant

Formula: C₉H₇ClF₃NO
Melting Pt: 99…101 °C
MDL Number: MFCD00018893
CAS Number: 3792-04-9

Order Now

Specification Test Results

Melting point 92-94°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR