2-(Pyridin-4-yl)acetic acid hydrochloride ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Pyridylacetic acid hydrochloride

002909-5G 002909-25G
101784-896EA 64.62 USD
101784-896 101784-898
2-(Pyridin-4-yl)acetic acid hydrochloride ≥98%
2-(Pyridin-4-yl)acetic acid hydrochloride

TCA listed: T
Hazard: Irritant

Formula: C₇H₈ClNO₂
MW: 173.60 g/mol
Melting Pt: 141 °C (dec.)
MDL Number: MFCD00012827
CAS Number: 6622-91-9

Order Now

Specification Test Results

Melting point 141°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR