Methyl-2-[2-(trifluoromethyl)quinolin-4-ylthio]acetate

Supplier: Matrix Scientific

002898-1G 002898-5G
101784-878EA 95.38 USD
101784-878 101784-880
Methyl-2-[2-(trifluoromethyl)quinolin-4-ylthio]acetate
Methyl-2-[2-(trifluoromethyl)quinolin-4-ylthio]acetate

Hazard: Irritant

Formula: C₁₃H₁₀F₃NO₂S
MW: 301.29 g/mol
Melting Pt: 96…98 °C
MDL Number: MFCD00219657
CAS Number: 175203-42-6

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR