Phloretin ≥98%

Supplier: Matrix Scientific

002894-250MG 002894-1G
101784-874EA 15.38 USD
101784-874 101784-876
Phloretin ≥98%
Phloretin

Hazard: Irritant

Formula: C₁₅H₁₄O₅
MW: 274.28 g/mol
Melting Pt: 260…262 °C (dec.)
MDL Number: MFCD00002288
CAS Number: 60-82-2

Order Now

Specification Test Results

Melting point 260-262°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR