3-(Trifluoromethoxy)cinnamic acid ≥96%

Supplier: Matrix Scientific

Synonyms: 3-(3-(Trifluoromethoxy)phenyl)acrylic acid

002883-5G 002883-25G
101784-844EA 72.31 USD
101784-844 101784-846
3-(Trifluoromethoxy)cinnamic acid ≥96%
3-(Trifluoromethoxy)cinnamic acid

Hazard: Irritant

Formula: C₁₀H₇F₃O₃
MW: 232.16 g/mol
Melting Pt: 92-95°C
MDL Number: MFCD00066337
CAS Number: 168833-80-5

Order Now

Specification Test Results

Melting point 92-95°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR