6-(2,2,2-Trifluoroethoxy)pyridine-3-thio-carboxamide
Supplier: Matrix Scientific
Synonyms:
6-(2,2,2-Trifluoroethoxy)thionicotinamide
002880-100MG
002880-250MG
101784-832EA
36.92
USD
101784-832
101784-834
6-(2,2,2-Trifluoroethoxy)pyridine-3-thio-carboxamide
6-(2,2,2-Trifluoroethoxy)pyridine-3-thio-carboxamide
Hazard: Irritant
Formula:
C₈H₇F₃N₂OS MW: 236.22 g/mol Melting Pt: 120…123 °C |
MDL Number:
MFCD00277205 CAS Number: 175277-59-5 |
Specification Test Results
Melting point | 120-123°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...