tert-Butyl (piperidin-4-ylmethyl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Piperidylmethyl)carbamic acid tert-Butyl ester, tert-Butyl (4-Piperidylmethyl)carbamate, 4-(tert-Butoxycarbonylaminomethyl)piperidine, 4-(Boc-aminomethyl)piperidine

016373-1G 016373-5G
101804-610EA 27.69 USD
101804-610 101804-612
tert-Butyl (piperidin-4-ylmethyl)carbamate ≥95%
tert-Butyl (piperidin-4-ylmethyl)carbamate

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Melting Pt: 153 °C
MDL Number: MFCD01631214
CAS Number: 135632-53-0

Order Now

Specification Test Results

Melting point 153°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR