2-Bromo-4-fluorophenylacetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(2-Bromo-4-fluorophenyl)acetic acid

009436-25G 009436-100G
101797-114EA 63.08 USD
101797-114 101797-116
2-Bromo-4-fluorophenylacetic acid ≥98%
2-Bromo-4-fluorophenylacetic acid

Hazard: Irritant

Formula: C₈H₆BrFO₂
MW: 233.0300 g/mol
Melting Pt: 108-111°C
MDL Number: MFCD04107675
CAS Number: 61150-59-2

Order Now

Specification Test Results

Melting point 108-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR