6-Cyanonicotinic acid ≥95%

Supplier: Matrix Scientific

009428-250MG 009428-1G
101797-076EA 340 USD
101797-076 101797-078
6-Cyanonicotinic acid ≥95%
6-Cyanonicotinic acid

Hazard: Irritant/Irritant-Harmful

Formula: C₇H₄N₂O₂
Melting Pt: 198…201 °C
MDL Number: MFCD01318135
CAS Number: 70165-31-0

Order Now

Specification Test Results

Melting point 192-194°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR