2-{[1-(tert-Butoxycarbonyl)piperidin-4-yl]sulfonyl}benzoic acid

Supplier: Matrix Scientific

009379-250MG 009379-1G
101796-976EA 123.08 USD
101796-976 101796-978
2-{[1-(tert-Butoxycarbonyl)piperidin-4-yl]sulfonyl}benzoic acid
2-{[1-(tert-Butoxycarbonyl)piperidin-4-yl]sulfonyl}benzoic acid

Hazard: Irritant

Formula: C₁₇H₂₃NO₆S
MW: 369.44 g/mol
Melting Pt: 191…193 °C
MDL Number: MFCD04037974
CAS Number: 849035-97-8

Order Now

Specification Test Results

Melting point 191-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR