1,2-Difluoro-4-(methylsulfonyl)benzene ≥97%

Supplier: Matrix Scientific

009332-1G 009332-5G
101796-882EA 44.62 USD
101796-882 101796-884
1,2-Difluoro-4-(methylsulfonyl)benzene ≥97%
1,2-Difluoro-4-(methylsulfonyl)benzene

Hazard: Irritant

Formula: C₇H₆F₂O₂S
Melting Pt: 71 °C
MDL Number: MFCD04037934
CAS Number: 424792-57-4

Order Now

Specification Test Results

Melting point 71°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR