4-(tert-Butoxycarbonyl)benzoic acid

Supplier: Matrix Scientific

Synonyms: mono-(tert-Butyl) terephthalate

009281-1G 009281-5G
101796-774EA 124.62 USD
101796-774 102663-572
4-(tert-Butoxycarbonyl)benzoic acid
4-(tert-Butoxycarbonyl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Melting Pt: 209…212 °C (dec.)
MDL Number: MFCD04037901
CAS Number: 20576-82-3

Order Now

Specification Test Results

Melting point 209-212°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR