3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Methylamino)propanoic acid, N-BOC protected

009279-1G 009279-5G
101796-768EA 44.62 USD
101796-768 101796-770
3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid ≥95%
3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid

Hazard: Irritant

Formula: C₉H₁₇NO₄
Melting Pt: 59…62 °C
MDL Number: MFCD03490508
CAS Number: 124072-61-3

Order Now

Specification Test Results

Melting point 59-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR