Trifluoropyruvic acid monohydrate

Supplier: Matrix Scientific

Synonyms: 3,3,3-trifluoro-2-oxopropanoic acid hydrate, 2,2-Dihydroxy-3,3,3-trifluoropropanoic acid monohydrate

009220-1G 009220-5G
101796-654EA 223.08 USD
101796-654 101796-656
Trifluoropyruvic acid monohydrate
Trifluoropyruvic acid monohydrate

Hazard: Corrosive

Formula: C₃HF₃O₃·H₂O
MW: 160.049 g/mol
Melting Pt: 122…125 °C
MDL Number: MFCD04038370
CAS Number: 10321-14-9

Order Now

Specification Test Results

Melting point 122-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR