2-(Trifluoroacetyl)pyrrole ≥99%
Supplier: Matrix Scientific
009185-1G
009185-5G
009185-25G
101796-592EA
58.46
USD
101796-592
101796-594
101796-596
2-(Trifluoroacetyl)pyrrole ≥99%
2-(Trifluoroacetyl)pyrrole
Hazard: Irritant
Formula:
C₆H₄F₃NO MW: 163.100 g/mol Melting Pt: 46…47 °C |
MDL Number:
MFCD00141809 CAS Number: 2557-70-2 |
Specification Test Results
Melting point | 48-50°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...