Potassium perfluorobutane-1-sulfonate ≥98%

Supplier: Matrix Scientific

Synonyms: Nonafluoro-1-butanesulphonic acid potassium salt, Potassium Perfluoro-1-butanesulphonate, Potassium Nonafluoro-1-butanesulphonate, Perfluoro-1-butanesulphonic acid potassium salt, Potassium nonafluoro-1-butanesulphonate

009149-5G 009149-25G
101796-496EA 73.85 USD
101796-496 101796-498
Potassium perfluorobutane-1-sulfonate ≥98%
Potassium perfluorobutane-1-sulphonate

Hazard: Irritant

Formula: C₄F₉KO₃S
MW: 338.1899 g/mol
Melting Pt: 300 °C (dec.)
MDL Number: MFCD00007515
CAS Number: 29420-49-3

Order Now

Specification Test Results

Melting point 300°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR