Perfluorodecaneamide ≥97%

Supplier: Matrix Scientific

009093-1G 009093-5G
101796-324EA 76.92 USD
101796-324 101796-326
Perfluorodecaneamide ≥97%
Perfluorodecaneamide

Hazard: Irritant

Formula: C₁₀H₂F₁₉NO
Melting Pt: 161…163 °C
MDL Number: MFCD04038334
CAS Number: 307-40-4

Order Now

Specification Test Results

Melting point 161-163°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR