4-(Perfluorooctyl)-2-methyl-2-butanol ≥98%

Supplier: Matrix Scientific

009090-500MG 009090-2.5G
101796-316EA 133.85 USD
101796-316 101796-318
4-(Perfluorooctyl)-2-methyl-2-butanol ≥98%
4-(Perfluorooctyl)-2-methyl-2-butanol

Hazard: Irritant

Formula: C₁₃H₁₁F₁₇O
Melting Pt: 50…51 °C
MDL Number: MFCD03788339
CAS Number: 141183-94-0

Order Now

Specification Test Results

Melting point 50-51°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR