Pentafluorophenyl sulfide ≥98%

Supplier: Matrix Scientific

Synonyms: Decafluorodiphenyl sulphide

009083-1G 009083-5G
101796-298EA 100 USD
101796-298 101796-300
Pentafluorophenyl sulfide ≥98%
Pentafluorophenyl sulfide

Hazard: Irritant

Formula: C₁₂F₁₀S
MW: 366.18 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD00000293
CAS Number: 1043-50-1

Order Now

Specification Test Results

Melting point 86-88°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR