Bis(pyridine)iodonium tetrafluoroborate

Supplier: Matrix Scientific

009055-1G 009055-5G
101796-222EA 163.08 USD
101796-222 101796-224
Bis(pyridine)iodonium tetrafluoroborate
Bis(pyridine)iodonium tetrafluoroborate

Hazard: Moisture Sensitive/Store in Freezer

Formula: C₁₀H₁₀BF₄IN₂
Melting Pt: 159-161°C (dec)
MDL Number: MFCD03703393
CAS Number: 15656-28-7

Order Now

Specification Test Results

Melting point 159-161°C(d


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR