2-(2-Hydroxy-1,1,1,3,3,3-hexafluoropropyl)-1-naphthol ≥97%
Supplier: Matrix Scientific
009001-1G
009001-5G
101796-068EA
258.46
USD
101796-068
101796-070
2-(2-Hydroxy-1,1,1,3,3,3-hexafluoropropyl)-1-naphthol ≥97%
2-(2-Hydroxy-1,1,1,3,3,3-hexafluoropropyl)-1-naphthol
Hazard: Irritant
Formula:
C₁₃H₈F₆O₂ MW: 310.2 g/mol Melting Pt: 119…120 °C |
MDL Number:
MFCD06247736 CAS Number: 2092-87-7 |
Specification Test Results
Melting point | 119-120°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...