2-Hydrazino-4-(trifluoromethyl)pyrimidine ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Hydrazinyl-4-(trifluoromethyl)pyrimidine, [4-(Trifluoromethyl)pyrimidin-2-yl]hydrazine

009000-1G 009000-5G
101796-064EA 110.77 USD
101796-064 101796-066
2-Hydrazino-4-(trifluoromethyl)pyrimidine ≥99%
2-Hydrazino-4-(trifluoromethyl)pyrimidine

Hazard: Irritant/Irritant-Harmful

Formula: C₅H₅F₃N₄
MW: 178.118 g/mol
Melting Pt: 99…101 °C
MDL Number: MFCD00115112
CAS Number: 197305-97-8

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR