Heptafluorobutyrylamidine ≥97%

Supplier: Matrix Scientific

008978-1G 008978-5G
101795-998EA 107.69 USD
101795-998 101796-000
Heptafluorobutyrylamidine ≥97%
Heptafluorobutyrylamidine

Hazard: Irritant

Formula: C₄H₃F₇N₂
Melting Pt: 49…51 °C
MDL Number: MFCD00236736
CAS Number: 375-19-9

Order Now

Specification Test Results

Melting point 49-51°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR