3-Fluoro-DL-tyrosine ≥97%

Supplier: Matrix Scientific

Synonyms: H-DL-Tyr(3-F)-OH, 2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid

008975-500MG 008975-2.5G
101795-986EA 176.92 USD
101795-986 101795-988
3-Fluoro-DL-tyrosine ≥97%
3-Fluoro-DL-tyrosine

Hazard: Irritant

Formula: C₉H₁₀FNO₃
MW: 199.18 g/mol
Melting Pt: 276…277 °C°C
MDL Number: MFCD00063075
CAS Number: 403-90-7

Order Now

Specification Test Results

Melting point 280°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR