(2-Fluorophenyl)glycine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Amino-2-(2-fluorophenyl)acetic acid

008971-1G 008971-5G
101795-980EA 36.92 USD
101795-980 101795-982
(2-Fluorophenyl)glycine ≥98%
(2-Fluorophenyl)glycine

Hazard: Irritant

Formula: C₈H₈FNO₂
MW: 169.16 g/mol
Melting Pt: 290 °C
MDL Number: MFCD00042726
CAS Number: 84145-28-8

Order Now

Specification Test Results

Melting point 290°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR